Assay Details
| Gene Symbol: LOC106998741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139986.2 | XP_014995472.1 | 1749 | 110 | ||
| GenBank mRNA | JU473067.1 | - | 821 | 110 | ||
More Information
Set Membership: |
|

LOC106998741
Genomic Map