Assay Details
| Gene Symbol: C15H9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002800088.3 | XP_002800134.2 | 3286 | 77 | ||
| GenBank mRNA | JV457386.1 | - | 2601 | 77 | ||
| JV707256.1 | - | 3018 | 77 | |||
More Information
Set Membership: |
|

C15H9orf131
Genomic Map