Assay Details
Gene Symbol: LOC705696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125931.2 | XP_014981417.1 | 2682 | 91 | ||
XM_015125932.2 | XP_014981418.1 | 2652 | 91 | |||
XM_028841348.1 | XP_028697181.1 | 2603 | 91 | |||
GenBank mRNA | JU320447.1 | - | 2609 | 91 | ||
JV044047.1 | - | 2528 | 91 | |||
JV524154.1 | - | 2622 | 91 | |||
JV725930.1 | - | 718 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
