Assay Details
| Gene Symbol: C6H5orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140057.2 | XP_014995543.2 | 919 | 82 | ||
| GenBank mRNA | JU325000.1 | - | 621 | 82 | ||
| JV712858.1 | - | 122 | 82 | |||
More Information
Set Membership: |
|

C6H5orf64
Genomic Map