Assay Details
| Gene Symbol: DDX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109944.2 | XP_014965430.2 | 1831 | 85 | ||
| GenBank mRNA | JU336941.1 | - | 1751 | 85 | ||
| JU477673.1 | - | 1756 | 85 | |||
| JV489178.1 | - | 1725 | 85 | |||
| JV726114.1 | - | 1755 | 85 | |||
| JV726115.1 | - | 1755 | 85 | |||
More Information
Set Membership: |
|

DDX18
Genomic Map