Assay Details
| Gene Symbol: GPR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091731.4 | XP_001091731.1 | 1397 | 87 | ||
| XM_028837254.1 | XP_028693087.1 | 1589 | 87 | |||
| XM_028837255.1 | XP_028693088.1 | 963 | 87 | |||
| GenBank mRNA | JU484786.1 | - | 627 | 87 | ||
More Information
Set Membership: |
|

GPR18
Genomic Map