Assay Details
| Gene Symbol: TINAGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836767.1 | XP_028692600.1 | 1389 | 58 | ||
| XM_028836771.1 | XP_028692604.1 | 1032 | 58 | |||
| GenBank mRNA | JV047909.1 | - | 1347 | 58 | ||
| JV047910.1 | - | 1347 | 58 | |||
| JV635562.1 | - | 1319 | 58 | |||
More Information
Set Membership: |
|

TINAGL1
Genomic Map