Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151419.2 | XP_015006905.1 | 1670 | 80 | ||
| GenBank mRNA | JV505163.1 | - | 283 | 80 | ||
| JV580447.1 | - | 283 | 80 | |||
More Information
Set Membership: |
|

SYT10
Genomic Map