Assay Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150239.2 | XP_015005725.2 | 1553 | 64 | ||
| GenBank mRNA | JU325611.1 | - | 1319 | 64 | ||
| JU493846.1 | - | 1190 | 64 | |||
| JV636385.1 | - | 1497 | 64 | |||
More Information
Set Membership: |
|

FOXRED2
Genomic Map