Assay Details
| Gene Symbol: C8H8orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145533.2 | XP_015001019.1 | 95 | |||
| GenBank mRNA | JV532756.1 | - | 121 | 95 | ||
| JV706421.1 | - | 147 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H8orf34| Gene Symbol: C8H8orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145533.2 | XP_015001019.1 | 95 | |||
| GenBank mRNA | JV532756.1 | - | 121 | 95 | ||
| JV706421.1 | - | 147 | 95 | |||