Assay Details
| Gene Symbol: INSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149580.2 | XP_015005066.2 | 1699 | 62 | ||
| GenBank mRNA | JU325871.1 | - | 323 | 62 | ||
| JU485968.1 | - | 759 | 62 | |||
More Information
Set Membership: |
|

INSM1
Genomic Map