Assay Details
| Gene Symbol: IRF2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126420.2 | XP_014981906.1 | 1045 | 83 | ||
| GenBank mRNA | JU327207.1 | - | 642 | 83 | ||
| JU475470.1 | - | 1009 | 83 | |||
| JV635457.1 | - | 1051 | 83 | |||
More Information
Set Membership: |
|

IRF2BP2
Genomic Map