Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123742.2 | XP_014979228.2 | 101 | 97 | ||
XM_015123743.2 | XP_014979229.2 | 101 | 97 | |||
XM_015123744.2 | XP_014979230.2 | 101 | 97 | |||
GenBank mRNA | JU320253.1 | - | 70 | 97 | ||
JU471202.1 | - | 101 | 97 | |||
JV660867.1 | - | 138 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123742.2, XM_015123743.2, XM_015123744.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
