Assay Details
| Gene Symbol: BTN3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135736.2 | XP_014991222.2 | 373 | 114 | ||
| XM_028847771.1 | XP_028703604.1 | 373 | 114 | |||
| XM_028847772.1 | XP_028703605.1 | 373 | 114 | |||
| GenBank mRNA | JU479348.1 | - | 428 | 114 | ||
More Information
Set Membership: |
|

BTN3A1
Genomic Map