Assay Details
| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082393.4 | XP_001082393.2 | 3981 | 69 | ||
| XM_015137552.2 | XP_014993038.2 | 3637 | 69 | |||
| GenBank mRNA | JV684103.1 | - | 223 | 69 | ||
More Information
Set Membership: |
|

MARCKS
Genomic Map