Assay Details
| Gene Symbol: LCMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106399.4 | XP_001106399.2 | 1733 | 100 | ||
| GenBank mRNA | JU328960.1 | - | 214 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCMT2| Gene Symbol: LCMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106399.4 | XP_001106399.2 | 1733 | 100 | ||
| GenBank mRNA | JU328960.1 | - | 214 | 100 | ||
Set Membership: |
|