Assay Details
| Gene Symbol: RAD9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113583.2 | XP_014969069.2 | 1246 | 82 | ||
| XM_015113584.2 | XP_014969070.2 | 2423 | 82 | |||
| XM_015113585.2 | XP_014969071.2 | 2377 | 82 | |||
| XM_015113586.2 | XP_014969072.2 | 1154 | 82 | |||
| XM_015113587.2 | XP_014969073.2 | 2333 | 82 | |||
| XM_015113589.2 | XP_014969075.2 | 2291 | 82 | |||
| XM_015113592.2 | XP_014969078.1 | 942 | 82 | |||
| XM_028833168.1 | XP_028689001.1 | 1839 | 82 | |||
| XM_028833169.1 | XP_028689002.1 | 1179 | 82 | |||
| XM_028833170.1 | XP_028689003.1 | 1138 | 82 | |||
| XM_028833171.1 | XP_028689004.1 | 2352 | 82 | |||
| XM_028833172.1 | XP_028689005.1 | 2123 | 82 | |||
| XM_028833173.1 | XP_028689006.1 | 901 | 82 | |||
| XM_028833174.1 | XP_028689007.1 | 2262 | 82 | |||
| GenBank mRNA | JU335738.1 | - | 952 | 82 | ||
| JU335739.1 | - | 952 | 82 | |||
| JU476628.1 | - | 963 | 82 | |||
| JV047815.1 | - | 962 | 82 | |||
| JV657718.1 | - | 925 | 82 | |||
More Information
Set Membership: |
|

RAD9A
Genomic Map