Assay Details
| Gene Symbol: CCL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126245.2 | XP_014981731.2 | 104 | 142 | ||
| GenBank mRNA | JU476729.1 | - | 114 | 142 | ||
| JU476730.1 | - | 114 | 142 | |||
| JU476731.1 | - | 114 | 142 | |||
More Information
Set Membership: |
|

CCL22
Genomic Map