Assay Details
| Gene Symbol: LOC700412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115498.2 | XP_014970984.2 | 2179 | 93 | ||
| GenBank mRNA | JU479184.1 | - | 618 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC700412| Gene Symbol: LOC700412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115498.2 | XP_014970984.2 | 2179 | 93 | ||
| GenBank mRNA | JU479184.1 | - | 618 | 93 | ||
Set Membership: |
|