Assay Details
| Gene Symbol: DMAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107783.4 | XP_001107783.3 | 611 | 71 | ||
| GenBank mRNA | JU350347.1 | - | 346 | 71 | ||
| JV632973.1 | - | 349 | 71 | |||
More Information
Set Membership: |
|

DMAC1
Genomic Map