Assay Details
| Gene Symbol: MRTFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125643.2 | XP_014981129.2 | 1315 | 107 | ||
| XM_015125644.2 | XP_014981130.2 | 1423 | 107 | |||
| XM_015125645.2 | XP_014981131.2 | 1296 | 107 | |||
| XM_015125646.2 | XP_014981132.2 | 1529 | 107 | |||
| XM_015125648.2 | XP_014981134.2 | 1260 | 107 | |||
| XM_015125649.2 | XP_014981135.2 | 3701 | 107 | |||
| GenBank mRNA | JU346229.1 | - | 1233 | 107 | ||
| JV719573.1 | - | 1236 | 107 | |||
More Information
Set Membership: |
|

MRTFB
Genomic Map