Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120460.2 | XP_014975946.2 | 1665 | 96 | ||
| XM_015120462.2 | XP_014975948.2 | 1389 | 96 | |||
| XM_015120463.2 | XP_014975949.2 | 1827 | 96 | |||
| XM_015120464.2 | XP_014975950.2 | 2049 | 96 | |||
| XM_015120465.2 | XP_014975951.1 | 804 | 96 | |||
| XM_015120466.2 | XP_014975952.1 | 1099 | 96 | |||
| XM_015120467.2 | XP_014975953.1 | 908 | 96 | |||
| XM_015120468.2 | XP_014975954.1 | 701 | 96 | |||
| XM_028836849.1 | XP_028692682.1 | 1134 | 96 | |||
| XM_028836850.1 | XP_028692683.1 | 639 | 96 | |||
| GenBank mRNA | JU322456.1 | - | 1392 | 96 | ||
| JV045986.1 | - | 1444 | 96 | |||
| JV045987.1 | - | 1449 | 96 | |||
| JV527780.1 | - | 1293 | 96 | |||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map