Assay Details
| Gene Symbol: OBP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834655.1 | XP_028690488.1 | 440 | 73 | ||
| GenBank mRNA | JV542832.1 | - | 310 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OBP2B| Gene Symbol: OBP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834655.1 | XP_028690488.1 | 440 | 73 | ||
| GenBank mRNA | JV542832.1 | - | 310 | 73 | ||
Set Membership: |
|