Assay Details
Gene Symbol: C14H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833319.1 | XP_028689152.1 | 671 | 98 | ||
XM_028833320.1 | XP_028689153.1 | 671 | 98 | |||
GenBank mRNA | JV626570.1 | - | 560 | 98 | ||
JV694088.1 | - | 571 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
