Assay Details
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092189.3 | XP_001092189.1 | 264 | 65 | ||
GenBank mRNA | JV075554.1 | - | 87 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
