Assay Details
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144776.2 | XP_015000262.2 | 536 | 108 | ||
| XM_028852360.1 | XP_028708193.1 | 799 | 108 | |||
| GenBank mRNA | JU339462.1 | - | 327 | 108 | ||
More Information
Set Membership: |
|

PRAG1
Genomic Map