Assay Details
| Gene Symbol: CAPN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112044.2 | XP_014967530.2 | 3359 | 88 | ||
| XM_015112045.2 | XP_014967531.2 | 3683 | 88 | |||
| XM_015112046.2 | XP_014967532.2 | 3242 | 88 | |||
| XM_015112047.2 | XP_014967533.2 | 3224 | 88 | |||
| XM_028831802.1 | XP_028687635.1 | 3284 | 88 | |||
| GenBank mRNA | JU343848.1 | - | 119 | 88 | ||
More Information
Set Membership: |
|

CAPN14
Genomic Map