Assay Details
| Gene Symbol: CKAP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092735.4 | XP_001092735.4 | 1822 | 96 | ||
| GenBank mRNA | JU481041.1 | - | 1706 | 96 | ||
| JV643331.1 | - | 540 | 96 | |||
More Information
Set Membership: |
|

CKAP2L
Genomic Map