Assay Details
| Gene Symbol: DLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144496.2 | XP_014999982.1 | 527 | 58 | ||
| GenBank mRNA | JV048586.1 | - | 317 | 58 | ||
| JV636437.1 | - | 229 | 58 | |||
More Information
Set Membership: |
|

DLK1
Genomic Map