Assay Details
| Gene Symbol: SOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121465.2 | XP_014976951.2 | 2123 | 84 | ||
| GenBank mRNA | JV721797.1 | - | 717 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX1| Gene Symbol: SOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121465.2 | XP_014976951.2 | 2123 | 84 | ||
| GenBank mRNA | JV721797.1 | - | 717 | 84 | ||
Set Membership: |
|