Assay Details
| Gene Symbol: SNX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148926.2 | XP_015004412.1 | 763 | 78 | ||
| GenBank mRNA | JV597668.1 | - | 545 | 78 | ||
| JV726330.1 | - | 394 | 78 | |||
| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV597667.1 | - | 545 | 78 | ||
More Information
Set Membership: |
|

SNX21
Genomic Map