Assay Details
| Gene Symbol: ORMDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151930.2 | XP_015007416.1 | 529 | 81 | ||
| GenBank mRNA | JU321296.1 | - | 408 | 81 | ||
| JU473223.1 | - | 390 | 81 | |||
More Information
Set Membership: |
|

ORMDL2
Genomic Map