Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134550.2 | XP_014990036.1 | 1421 | 90 | ||
| XM_028836407.1 | XP_028692240.1 | 766 | 90 | |||
| XM_028836408.1 | XP_028692241.1 | 941 | 90 | |||
| GenBank mRNA | JU321904.1 | - | 664 | 90 | ||
| JU471219.1 | - | 667 | 90 | |||
| JV045536.1 | - | 670 | 90 | |||
| JV566762.1 | - | 698 | 90 | |||
More Information
Set Membership: |
|

YARS1
Genomic Map