Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134550.2 | XP_014990036.1 | 1728 | 100 | ||
| XM_028836407.1 | XP_028692240.1 | 1073 | 100 | |||
| XM_028836408.1 | XP_028692241.1 | 1248 | 100 | |||
| GenBank mRNA | JU321904.1 | - | 971 | 100 | ||
| JU471219.1 | - | 974 | 100 | |||
| JV045536.1 | - | 977 | 100 | |||
| JV566762.1 | - | 1005 | 100 | |||
More Information
Set Membership: |
|

YARS1
Genomic Map