Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134550.2 | XP_014990036.1 | 1808 | 101 | ||
| XM_028836407.1 | XP_028692240.1 | 1153 | 101 | |||
| XM_028836408.1 | XP_028692241.1 | 1328 | 101 | |||
| GenBank mRNA | JU321904.1 | - | 1051 | 101 | ||
| JU471219.1 | - | 1054 | 101 | |||
| JV045536.1 | - | 1057 | 101 | |||
| JV566762.1 | - | 1085 | 101 | |||
More Information
Set Membership: |
|

YARS1
Genomic Map