Assay Details
| Gene Symbol: LOC708660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138769.2 | XP_014994255.1 | 1548 | 75 | ||
| GenBank mRNA | JU322996.1 | - | 1526 | 75 | ||
| JV709359.1 | - | 1532 | 75 | |||
More Information
Set Membership: |
|

LOC708660
Genomic Map