Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133903.2 | XP_014989389.2 | 1562 | 95 | ||
| GenBank mRNA | JU330178.1 | - | 116 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MACC1| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133903.2 | XP_014989389.2 | 1562 | 95 | ||
| GenBank mRNA | JU330178.1 | - | 116 | 95 | ||
Set Membership: |
|