Assay Details
| Gene Symbol: CCDC180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835047.1 | XP_028690880.1 | 5248 | 66 | ||
| GenBank mRNA | JU478462.1 | - | 370 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC180| Gene Symbol: CCDC180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835047.1 | XP_028690880.1 | 5248 | 66 | ||
| GenBank mRNA | JU478462.1 | - | 370 | 66 | ||
Set Membership: |
|