Assay Details
| Gene Symbol: HASPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090778.4 | XP_001090778.2 | 2243 | 89 | ||
| GenBank mRNA | JU494602.1 | - | 2119 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HASPIN| Gene Symbol: HASPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090778.4 | XP_001090778.2 | 2243 | 89 | ||
| GenBank mRNA | JU494602.1 | - | 2119 | 89 | ||
Set Membership: |
|