Assay Details
| Gene Symbol: KCNRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV049376.1 | - | 807 | 99 | ||
| JV704993.1 | - | 827 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNRG| Gene Symbol: KCNRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV049376.1 | - | 807 | 99 | ||
| JV704993.1 | - | 827 | 99 | |||
Set Membership: |
|