Assay Details
| Gene Symbol: PCYT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843530.1 | XP_028699363.1 | 1103 | 76 | ||
| XM_028843531.1 | XP_028699364.1 | 1030 | 76 | |||
| XM_028843532.1 | XP_028699365.1 | 1051 | 76 | |||
| XM_028843533.1 | XP_028699366.1 | 937 | 76 | |||
| XM_028843534.1 | XP_028699367.1 | 937 | 76 | |||
| XM_028843535.1 | XP_028699368.1 | 937 | 76 | |||
| GenBank mRNA | JU334852.1 | - | 918 | 76 | ||
| JU475877.1 | - | 909 | 76 | |||
| JV047278.1 | - | 922 | 76 | |||
| JV509061.1 | - | 863 | 76 | |||
More Information
Set Membership: |
|

PCYT1A
Genomic Map