Assay Details
| Gene Symbol: MRPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112795.2 | XP_014968281.1 | 362 | 75 | ||
| GenBank mRNA | JU334860.1 | - | 250 | 75 | ||
| JU334861.1 | - | 250 | 75 | |||
| JU475882.1 | - | 250 | 75 | |||
| JV047287.1 | - | 262 | 75 | |||
| JV453074.1 | - | 268 | 75 | |||
| JV703996.1 | - | 252 | 75 | |||
More Information
Set Membership: |
|

MRPL35
Genomic Map