Assay Details
| Gene Symbol: DMAC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143662.2 | XP_014999148.1 | 1078 | 146 | ||
| XM_015143663.2 | XP_014999149.1 | 1213 | 146 | |||
| XM_028851539.1 | XP_028707372.1 | 580 | 146 | |||
| GenBank mRNA | JU341844.1 | - | 495 | 146 | ||
| JV712101.1 | - | 493 | 146 | |||
More Information
Set Membership: |
|

DMAC2L
Genomic Map