Assay Details
| Gene Symbol: ATOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141269.2 | XP_014996755.1 | 2083 | 55 | ||
| GenBank mRNA | JU473629.1 | - | 94 | 55 | ||
| JU473630.1 | - | 94 | 55 | |||
| JV047157.1 | - | 49 | 55 | |||
| JV530023.1 | - | 73 | 55 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015141269.2 transcripts.
Set Membership: |
|

ATOX1
Genomic Map