Assay Details
Gene Symbol: LOC100425888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832942.1 | XP_028688775.1 | 550 | 119 |
Gene Symbol: LOC114669834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832952.1 | XP_028688785.1 | 486 | 119 |

More Information
Set Membership: |
![]() ![]() |
