Assay Details
| Gene Symbol: LOC698851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU333862.1 | - | 702 | 101 | ||
| JV577582.1 | - | 726 | 101 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC698851| Gene Symbol: LOC698851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU333862.1 | - | 702 | 101 | ||
| JV577582.1 | - | 726 | 101 | |||
Set Membership: |
|