Assay Details
| Gene Symbol: ZSCAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847750.1 | XP_028703583.1 | 554 | 97 | ||
| GenBank mRNA | JU337307.1 | - | 546 | 97 | ||
| JU470742.1 | - | 512 | 97 | |||
More Information
Set Membership: |
|

ZSCAN12
Genomic Map