Assay Details
| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107923.3 | XP_001107923.1 | 1050 | 88 | ||
| GenBank mRNA | JU341221.1 | - | 803 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPAR3| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107923.3 | XP_001107923.1 | 1050 | 88 | ||
| GenBank mRNA | JU341221.1 | - | 803 | 88 | ||
Set Membership: |
|