Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126009.2 | XP_014981495.2 | 422 | 95 | ||
| GenBank mRNA | JU477156.1 | - | 407 | 95 | ||
| JV045678.1 | - | 281 | 95 | |||
| JV593256.1 | - | 357 | 95 | |||
| JV661119.1 | - | 409 | 95 | |||
More Information
Set Membership: |
|

VKORC1
Genomic Map