Assay Details
| Gene Symbol: ITPRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU495357.1 | - | 2420 | 114 | ||
| JV664143.1 | - | 2416 | 114 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPRIP| Gene Symbol: ITPRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU495357.1 | - | 2420 | 114 | ||
| JV664143.1 | - | 2416 | 114 | |||
Set Membership: |
|